missing translation for 'onlineSavingsMsg'
Learn More

LRIT2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA558105

Additional Details : Weight : 0.02000kg

Product Code. 15755991

  • £427.00 / 100µL
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: LVISLHVQPA QALHAPDSLS IPSEGNAYID LRVVKQTVHG ILLEWLAVAD TSKEEWFTLY IASDEAFRKE VVHIGPGINT YAVDDLLPGT KYE Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.

LRIT2 Polyclonal antibody specifically detects LRIT2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

LRIT2
Polyclonal
Unconjugated
LRIT2
leucine rich repeat containing 22; leucine rich repeat, Ig-like and transmembrane domains 2; leucine-rich repeat, Ig-like and transmembrane domains 2; leucine-rich repeat, immunoglobulin-like and transmembrane domains 2; leucine-rich repeat, immunoglobulin-like domain and transmembrane domain-containing protein 2; Leucine-rich repeat-containing protein 22; LRIT2; LRRC22
Rabbit
Antigen affinity chromatography
RUO
340745
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A6NDA9
LRIT2
Recombinant protein corresponding to Human LRIT2
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only