missing translation for 'onlineSavingsMsg'
Learn More

LP 44, Tocris Bioscience™

High affinity 5-HT7 agonist

Brand:  Tocris (Bio-Techne) 253410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461414

  • £183.48 / 10mg


Estimated Shipment
03-01-2019
Add to basket

Description

Description

High affinity 5-HT7 receptor agonist (Ki = 0.22 nM) that displays selectivity over 5-HT1A and 5-HT2A receptors (200- and > 1000-fold respectively). Induces relaxation of substance P-stimulated guinea pig ileum (EC50 = 2.56 μM).
Specifications

Chemical Identifiers

824958-12-5
488.131
4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphth alenyl-1-piperazinehexanamide hydrochloride, 4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphthalenyl-1-piperazinehexanamide hydrochloride, lp 44 hydrochloride, lp hplc , solid, 4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphthalenyl-1-piperazinehexanamidehydrochloride, 6-4-2-methylthio phenyl piperazin-1-yl-n-1,2,3,4-tetrahydronaphthalen-1-yl hexanamide hydrochloride
6-[4-(2-methylsulfanylphenyl)piperazin-1-yl]-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide;hydrochloride
C27H38ClN3OS
DWGKCWWWKHCVDH-UHFFFAOYSA-N
11225543
CSC1=CC=CC=C1N2CCN(CC2)CCCCCC(=O)NC3CCCC4=CC=CC=C34.Cl

Specifications

LP 44
824958-12-5
4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphth alenyl-1-piperazinehexanamide hydrochloride, 4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphthalenyl-1-piperazinehexanamide hydrochloride, lp 44 hydrochloride, lp hplc , solid, 4-2-methylthio phenyl-n-1,2,3,4-tetrahydro-1-naphthalenyl-1-piperazinehexanamidehydrochloride, 6-4-2-methylthio phenyl piperazin-1-yl-n-1,2,3,4-tetrahydronaphthalen-1-yl hexanamide hydrochloride
CSC1=CC=CC=C1N2CCN(CC2)CCCCCC(=O)NC3CCCC4=CC=CC=C34.Cl
488.131
488.13
10mg
C27H38ClN3OS
DWGKCWWWKHCVDH-UHFFFAOYSA-N
6-[4-(2-methylsulfanylphenyl)piperazin-1-yl]-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide;hydrochloride
11225543
>98%
Videos