missing translation for 'onlineSavingsMsg'
Learn More

LP 12 hydrochloride, Tocris Bioscience™

5-HT7 agonist

Brand:  Tocris (Bio-Techne) 292510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462054

  • £178.20 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

5-HT7 receptor agonist displaying selectivity over D2, 5-HT1A and 5-HT2A receptors (Ki values are 0.22, 7.3, 52.7 and 326 nM respectively). Induces relaxation of substance P-induced contractions in guinea pig ileum (EC50 = 1.77 μM).
Specifications

Chemical Identifiers

1185136-22-4
518.142
lp 12 hydrochloride, lp 12 hydrochloride hydrate, 4-2-diphenyl-n-1,2,3,4-tetrahydronaphthalen-1-yl-1-piperazinehexanamide hydrochloride
6-[4-(2-phenylphenyl)piperazin-1-yl]-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide;hydrochloride
C32H40ClN3O
XNDUOTNYUWDFKR-UHFFFAOYSA-N
56972187
C1CC(C2=CC=CC=C2C1)NC(=O)CCCCCN3CCN(CC3)C4=CC=CC=C4C5=CC=CC=C5.Cl

Specifications

LP 12 hydrochloride
1185136-22-4
lp 12 hydrochloride, lp 12 hydrochloride hydrate, 4-2-diphenyl-n-1,2,3,4-tetrahydronaphthalen-1-yl-1-piperazinehexanamide hydrochloride
C1CC(C2=CC=CC=C2C1)NC(=O)CCCCCN3CCN(CC3)C4=CC=CC=C4C5=CC=CC=C5.Cl
518.142
518.13
10mg
C32H40ClN3O
XNDUOTNYUWDFKR-UHFFFAOYSA-N
6-[4-(2-phenylphenyl)piperazin-1-yl]-N-(1,2,3,4-tetrahydronaphthalen-1-yl)hexanamide;hydrochloride
56972187
>99%
Videos