missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LOX-1/OLR1 Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | LOX-1/OLR1 |
---|---|
Clone | 214001 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Description
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
LOX-1/OLR1 | |
LYOPH | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
AAG44998 | |
4973 | |
Mouse myeloma cell line NS0-derived recombinant mouse LOX-1/OLR1 Arg60-Ile363 Accession # AAG44998 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse LOX-1/OLR1 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human LOX-1 or human LDL, HDL or VLDL is observed. |
214001 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
OLR1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only