missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Low-Temperature Heated Circulating Baths, TXF200 Series

Brand:  Grant Instruments™ TXF200-R3

Code : 57

Additional Details : Weight : 24.00000kg

Product Code. 12717849

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

-30 to 100°C
230, 120V
230V, 2kW, 50/60Hz and 120V, 1.5kW, 50/60Hz
Stainless Steel
5L
50/60Hz
22L/min.
530mbar
Videos
Documentation