Learn More
Thermo Scientific™ Low Profile Strip Tubes without Caps
Increase the efficiency of reactions with these low-profile strip tubes for PCR and qPCR applications.
Brand:  Thermo Scientific™ AB-0848
Code : 04
Additional Details : Weight : 0.26000kg
 View more versions of this product
Product Code. 11524804
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 µL.
  • Suitable for 0.2 mL thermal cycler blocks
  • 150 µL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A–H end tabs
  • Ultra Clear caps are ideal for qPCR
PCR, qPCR and Real-Time PCR
Certified free from human DNA, Rnases, and Dnases.
Strips of 12; 10 Packs of 8
Certified free of human DNA, RNase, DNase, and endotoxins
ABgene Low Profile Strip Tubes