missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Low Form Inconel Crucible

Brand:  Alfa Aesar™ 035949.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15413605

  • £45.79 / Each


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Specifications

35mL
38mm
30mm
Inconel
Low Form
46mm
Inconel Crucible
Videos
SDS