missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: DEHA Solution

Brand:  Lovibond™ 461181

Code : A0

Additional Details : CAS Number : 7697-37-2 Weight : 0.10000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

 View more versions of this product

Product Code. 11739548

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Yellow
Nitric Acid 2.5 to 5%
Liquid
Colorimeter and spectrophotometer
20°C
DEHA Reagent Solution
0 to 0.5mg/L
Water analysis
0.1
100mL
Videos
Documentation

Technical Documents