Learn More
Lovibond™ PD250 Reagent Dispenser Refill: DPD Free Chlorine and Total Chlorine
For use with SC450 Chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
Brand:  Lovibond™ 530160
Code : NEW
Additional Details : Weight : 0.30000kg
Product Code. 15301715
  • £77.70 / Pack of 2
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Specifications
For chlorine testing
Includes 1 vial of DPD free chlorine reagent and 1 vial of DPD total chlorine reagent
Replacement Free and Total Chlorine Reagent
Used with SC450 chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
250 Tests