missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Colorimeter Accessories for AF710-3 Tintometer

Brand:  Lovibond™ 122340

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11857474

  • £13.65 / Each
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For use with Lovibond Colorimeter (09-053-362).

Accessories:

  • Sample Tubes: pair of glass sample tubes to AOCS requirements
  • Tunsten Halogen Lamp: 12V 20w
  • Drip Tray: For use with porcelain (09-053-366)
  • Porcelain White Reference: for AOCS; measures 85 x 170mm

Specifications

Specifications

Tungsten Halogen Lamp
Lovibond AF710-3 Tintometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers