missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits
Accurate test kits for routine water analysis
£54.73 - £64.75
Specifications
For Use With (Application) | For routine water analysis |
---|---|
Sufficient For | 100 Tests |
Product Code | Brand | Type | Range | Sufficient For | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Sufficient For | Price | Quantity & Availability | |||
10356762
|
Lovibond™
147430 |
Copper | 0-5.0mg/L | 100 Tests |
£64.75
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||
10759674
|
Lovibond™
147280 |
Bromine Color Disc | 0 to 5mg/L | 100 Tests |
£54.73
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||
11715528
|
Lovibond™
147430 |
Copper | 0-5.0mg/L | 100 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
- Color discs and reagents are available separately
Specifications
For routine water analysis | |
100 Tests |