missing translation for 'onlineSavingsMsg'
Learn More

Loreclezole hydrochloride, Tocris Bioscience™

Subtype-selective GABAA modulator

Brand:  Tocris (Bio-Techne) 1295/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13449073

  • £189.63 / 10mg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Subtype-selective GABAA receptor modulator. Acts as a positive allosteric modulator of β2 or β3-subunit containing receptors. Also acts as a negative modulator at a novel regulatory site, enhancing GABAA receptor sensitization. Inhibits homomeric ρ1 GABAC receptors.
Specifications

Chemical Identifiers

117857-45-1
310.987
loreclezole hydrochloride, dsstox_cid_25676, dsstox_rid_81050, dsstox_gsid_45676, cas-nocas_45676, z-1-2-chloro-2-2,4-dichlorophenyl vinyl-1h-1,2,4-triazole hydrochloride, z-1-2-chloro-2-2,4-dichlorophenyl ethenyl-1h-1,2,4-triazole hydrochloride
1-[(Z)-2-chloro-2-(2,4-dichlorophenyl)ethenyl]-1,2,4-triazole;hydrochloride
C10H7Cl4N3
AWKSVFLKVCVVFQ-MDZFRNKHSA-N
45073437
C1=CC(=C(C=C1Cl)Cl)C(=CN2C=NC=N2)Cl.Cl

Specifications

10mg
117857-45-1
loreclezole hydrochloride, dsstox_cid_25676, dsstox_rid_81050, dsstox_gsid_45676, cas-nocas_45676, z-1-2-chloro-2-2,4-dichlorophenyl vinyl-1h-1,2,4-triazole hydrochloride, z-1-2-chloro-2-2,4-dichlorophenyl ethenyl-1h-1,2,4-triazole hydrochloride
C1=CC(=C(C=C1Cl)Cl)C(=CN2C=NC=N2)Cl.Cl
310.987
311
Loreclezole hydrochloride
C10H7Cl4N3
AWKSVFLKVCVVFQ-MDZFRNKHSA-N
1-[(Z)-2-chloro-2-(2,4-dichlorophenyl)ethenyl]-1,2,4-triazole;hydrochloride
45073437
>99%
Videos
Documents