missing translation for 'onlineSavingsMsg'
Learn More

LOE 908 hydrochloride, Tocris Bioscience™

Broad spectrum cation channel blocker; neuroprotective

Brand:  Tocris (Bio-Techne) 231450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431014

  • £658.56 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Broad spectrum cation channel blocker. Inhibits several cation channels including store-operated calcium channels (SOCs), voltage-operated calcium channels (VOCs), non-selective cation channels (NSCCs), AMPA, NMDA, Na+ and K+ channels. Neuroprotective; reduces cortical infarct size and improves neurological outcome following middle cerebral artery occlusion.
Specifications

Chemical Identifiers

143482-60-4
749.298
loe 908 hydrochloride, pinokalant hydrochloride, dsstox_cid_26305, dsstox_rid_81526, dsstox_gsid_46305, 2-6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl-2-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl acetamide hydrochloride, 3,4-dihydro-6,7-dimethoxy-?-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl-1-isoquinolineacetamide hydrochloride
2-(6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl)-2-phenyl-N,N-bis[2-(2,3,4-trimethoxyphenyl)ethyl]acetamide;hydrochloride
C41H49ClN2O9
UDKSNBMNKLDHNK-UHFFFAOYSA-N
56846451
COC1=C(C(=C(C=C1)CCN(CCC2=C(C(=C(C=C2)OC)OC)OC)C(=O)C(C3=CC=CC=C3)C4=NCCC5=CC(=C(C=C54)OC)OC)OC)OC.Cl

Specifications

50mg
143482-60-4
loe 908 hydrochloride, pinokalant hydrochloride, dsstox_cid_26305, dsstox_rid_81526, dsstox_gsid_46305, 2-6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl-2-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl acetamide hydrochloride, 3,4-dihydro-6,7-dimethoxy-?-phenyl-n,n-bis 2-2,3,4-trimethoxyphenyl ethyl-1-isoquinolineacetamide hydrochloride
COC1=C(C(=C(C=C1)CCN(CCC2=C(C(=C(C=C2)OC)OC)OC)C(=O)C(C3=CC=CC=C3)C4=NCCC5=CC(=C(C=C54)OC)OC)OC)OC.Cl
749.298
749.29
LOE 908 hydrochloride
C41H49ClN2O9
UDKSNBMNKLDHNK-UHFFFAOYSA-N
2-(6,7-dimethoxy-3,4-dihydroisoquinolin-1-yl)-2-phenyl-N,N-bis[2-(2,3,4-trimethoxyphenyl)ethyl]acetamide;hydrochloride
56846451
>98%
Videos
Documents