missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Locostatin, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
50 mg
Description
Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Locostatin is a cell-permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration
Specifications
Color | White |
Quantity | 10 mg |
Formula Weight | 245.27 |
Physical Form | Powder |
Chemical Name or Material | Locostatin |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'