missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Locker, perforated door, grey/blue, two tier, 1800mm x 300mm x 300mm

Brand:  Key Industrial Equipment™ A330153

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12885418

  • £206.65 / Each

Estimated Shipment date
from Supplier 25-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Quality steel lockers with perforated doors and BioCote antibacterial technology.
  • Two tier
  • Blue
  • Dimensions (H x W x D): 1800mm x 300mm x 300mm
Videos
Documents