missing translation for 'onlineSavingsMsg'
Learn More

LMO2, clone: 1A9-3B11, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9899-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218227

  • £188.13 / 100µg


Estimated Shipment
05-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This 1A9-3B11 monoclonal antibody reacts with human LMO2, a member of the LIM-only family of zinc finger proteins. LMO2 possesses two LIM domains for homodimerization and an N-terminal transactivation domain. This 24-kDa transcription factor can be found as part of a transcriptional complex composed of TAL1, E47, GATA-1, and LDB1 proteins. LMO2 is expressed in myeloid cells, germinal center B cells, vascular endothelium, and a variety of cancers, including diffuse large B cell lymphomas (DLBCL), T cell acute lymphoblastic lymphomas, and vascular neoplasms. Expression of LMO2 in germinal center B cells has been shown to be regulated by microRNAs, specifically hsa-miR-223. Although the function of LMO2 remains unclear in germinal center B cells, this factor is involved in hematopoiesis and oncogenesis. In addition, expression of this molecule has been demonstrated to be prognostic marker for survival in DLBCL patients.Applications Reported: This 1A9-3B11 antibody has been reported for use in immunohistochemical staining.Applications Tested: This 1A9-3B11 antibody has been tested by immunohistochemistry on human tonsil tissue and western blot on the Raji cell line. This can be used at 0.5-10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

LMO2
1A9-3B11
Unconjugated
Purified
IgG1
RUO
Primary
Human
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C.
Monoclonal
Videos