Learn More
Invitrogen™ LIVE/DEAD™ Fixable Far Red Dead Cell Stain Kit, for 633 or 635 nm excitation
The LIVE/DEAD™ Fixable Far-Red Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.
Brand: Invitrogen™ L10120
183.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
- Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
- Kit has been optimized and validated for use with a red laser flow cytometer
- Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
- Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
- If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
- LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs
Optimal brightness
- The LIVE/DEAD™ Fixable Far-Red Stain was selected based on its fluorescent properties to provide a bright signal when excited with a red laser
- The far-red fluorescent reactive dye has an excitation maximum of ∼633 nm making it ideal for use with the red or HeNe laser with an emission of ∼655 nm
- Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments
Specifications
Impermeant | |
Far Red | |
LIVE/DEAD™ Fixable Far Red Dead Cell Stain Kit, for 633 or 635 nm excitation | |
Viability Assay | |
Stain | |
655 nm | |
Solid | |
LIVE/DEAD™ | |
Room Temperature |
Eukaryotic Cells | |
Contains 5 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO. Store at -20°C. |
|
Fluorescence | |
Flow Cytometer | |
LIVE/DEAD™ Fixable Far Red Dead Cell Stain | |
633 nm | |
Tube(s) | |
200 Assays | |
DMSO (Dimethylsulfoxide) |
For Research Use Only. Not for use in diagnostic procedures.