missing translation for 'onlineSavingsMsg'
Learn More

Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride, 0.5M solution in THF, AcroSeal™, Thermo Scientific™

Catalog Number p-4229003
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
79172-99-9, 109-99-9
C21H46AlLiO3
380.52
MFCD00012222
FXBRQZDWJRAIEB-UHFFFAOYSA-N
lithium tris 3-ethyl-3-pentyl oxy aluminohydride
16689417
[Li+].CCC(CC)(CC)O[AlH-](OC(CC)(CC)CC)OC(CC)(CC)CC
This item is not returnable. View return policy
79172-99-9, 109-99-9
C21H46AlLiO3
380.52
MFCD00012222
FXBRQZDWJRAIEB-UHFFFAOYSA-N
lithium tris 3-ethyl-3-pentyl oxy aluminohydride
16689417
[Li+].CCC(CC)(CC)O[AlH-](OC(CC)(CC)CC)OC(CC)(CC)CC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Solution
Quantity 100 mL
Flash Point −17°C
IUPAC Name lithium(1+) tris[(3-ethylpentan-3-yl)oxy]alumanuide
Formula Weight 380.51
Concentration or Composition (by Analyte or Components) 0.40 to 0.60M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.904
Density 0.9040g/mL
Chemical Name or Material Lithium tris[(3-ethyl-3-pentyl)oxy]aluminohydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lithium tris¢(3-ethyl-3-pentyl)oxy!aluminohydride, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'