missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium triethylborohydride, 1M in THF, packaged under Argon in resealable ChemSeal™ bottles

Brand:  Alfa Aesar™ 042491.AP

Code : NEW

Additional Details : CAS Number : 22560-16-3 Weight : 0.01000kg Transport : UN number : 3399 Chem class : 1 Pack group : I

Quantity 500mL

 View more versions of this product

Product Code. 11306109

  • £304.90 / 500mL


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Lithium triethylborohydride
LiB(CH2CH3)3H
MFCD00011703
4151240
Lithium triethylhydridoborate
22560-16-3
105.94
UN3399
14,5544

Specifications

Lithium triethylborohydride
ChemSeal™ bottles
LiB(CH2CH3)3H
0.892g/mL
UN3399
14,5544
1M in THF
500mL
Moisture Sensitive
22560-16-3
Liquid
105.94
MFCD00011703
4151240
Lithium triethylhydridoborate
Colorless
−17°C (1°F)
Videos
SDS