missing translation for 'onlineSavingsMsg'
Learn More

Lithium triethylborohydride, 1M in THF, Thermo Scientific™

Catalog Number p-7080497
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
22560-16-3
C6H16BLi
105.94
MFCD00011703
WCJAYABJWDIZAJ-UHFFFAOYSA-N
Lithium triethylhydridoborate
lithium(1+) triethylboranuide
[Li+].CC[BH-](CC)CC
This item is not returnable. View return policy
22560-16-3
C6H16BLi
105.94
MFCD00011703
WCJAYABJWDIZAJ-UHFFFAOYSA-N
Lithium triethylhydridoborate
lithium(1+) triethylboranuide
[Li+].CC[BH-](CC)CC

As reducing agent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.892g/mL
Flash Point −17°C (1°F)
Linear Formula LiB(CH2CH3)3H
Quantity 25 mL
UN Number UN3399
Beilstein 4151240
Sensitivity Moisture Sensitive
Merck Index 14,5544
Formula Weight 105.94
Concentration or Composition (by Analyte or Components) 1M in THF
Physical Form Liquid
Chemical Name or Material Lithium triethylborohydride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'