missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-tert-butoxyaluminohydride, 93-98%, Thermo Scientific Chemicals

Catalog Number 10570512
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
17476-04-9
C12H28AlLiO3
254.27
MFCD00011532
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
17476-04-9
C12H28AlLiO3
254.27
MFCD00011532

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 319.0°C
Color Gray-White to White
Density 0.9
Infrared Spectrum Authentic
Assay Percent Range 93% min
Packaging Glass Bottle packed in tin can
Linear Formula LiAI[OC(CH3)3]3H
Quantity 100 g
Fieser 01,620; 02,251; 03,188; 04,312; 10,248; 12,286; 13,342; 16,197
Solubility Information Solubility in water: reacts violently. Other solubilities: 36g/100g thf (25°C),2g/100g diethyl ether (25°C),41g/100g diglyme (25°C)
Sublimation Point 280 °C
Formula Weight 254.27
Percent Purity 93 to 98%
Physical Form Powder
Chemical Name or Material Lithium tri-tert-butoxyaluminohydride, 93-98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium tri-tert-butoxyaluminohydride
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'