missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-sec-butylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 176451000

Code : 29

Additional Details : CAS Number : 38721-52-7 Weight : 0.14000kg Transport : UN number : 3129 Chem class : 4.3 Pack group : I


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

 View more versions of this product

Product Code. 10144290

  • £31.60 / 100mL


EU Stock 9
Add to basket

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Description

Description

Reduction reagent
Specifications

Chemical Identifiers

Lithium tri-sec-butylborohydride
C12H28BLi
190.11
Lithium triisobutylhydroborate, Lithium triisobutylhydroborate
1113-78-6
LiB[CH(CH3)C2H5]3H
MFCD00011708

Specifications

Lithium tri-sec-butylborohydride
AcroSeal™ Glass Bottle in Tin Can
C12H28BLi
190.11
MFCD00011708
15, 8564
Clear colorless
1113-78-6
Liquid
LiB[CH(CH3)C2H5]3H
0.8900g/mL
04,312; 06,348; 07,307; 08,308; 10,248; 12,286; 14,195; 15,192
Lithium triisobutylhydroborate, Lithium triisobutylhydroborate
100mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lithium tri-sec-butylborohydride, 1M solution in THF

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS