missing translation for 'onlineSavingsMsg'
Learn More

Lithium tri-sec-butylborohydride, 1.0M solution in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-7058689
Click to view available options
Quantity:
25 mL
500 mL
This item is not returnable. View return policy
38721-52-7
C12H28BLi
190.11
MFCD00011708
ACJKNTZKEFMEAK-UHFFFAOYNA-N
lithium tri-sec-butylhydroborate, l-selectridesolution, lithium tri-sec-butylborohydride solution, l-selectride™ solution, lithium tri butan-2-yl boron 1-, lithium tri-sec-butylborohydride l-selectride , 1m in thf, lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal, lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
lithium(1+) tris(butan-2-yl)boranuide
[Li+].CCC(C)[BH-](C(C)CC)C(C)CC
This item is not returnable. View return policy
38721-52-7
C12H28BLi
190.11
MFCD00011708
ACJKNTZKEFMEAK-UHFFFAOYNA-N
lithium tri-sec-butylhydroborate, l-selectridesolution, lithium tri-sec-butylborohydride solution, l-selectride™ solution, lithium tri butan-2-yl boron 1-, lithium tri-sec-butylborohydride l-selectride , 1m in thf, lithium tri-sec-butylborohydride 1.0 m solution in thf, spcseal, lithium tri-sec-butylborohydride, 1.0m solution in thf, packaged under argon in resealable chemseal bottles
lithium(1+) tris(butan-2-yl)boranuide
[Li+].CCC(C)[BH-](C(C)CC)C(C)CC

As a reducing agent of olefinic groups and ketones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.870g/mL
Flash Point −17°C (1°F)
Packaging ChemSeal™ bottles
Linear Formula LiB[CH(CH3)CH2CH3]3H
Quantity 25 mL
UN Number UN3394
Beilstein 4007327
Sensitivity Moisture Sensitive
Merck Index 14,8427
Formula Weight 190.11
Concentration or Composition (by Analyte or Components) 1.0M solution in THF
Physical Form Liquid
Chemical Name or Material Lithium tri-sec-butylborohydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'