missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lithium titanium oxide, 99% (metals basis)

Brand:  Alfa Aesar™ 041650.30

Code : NEW

Additional Details : CAS Number : 12031-82-2 Weight : 0.25000kg

Quantity 250g

 View more versions of this product

Product Code. 11388788

  • £135.04 / 250g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

Lithium titanium oxide
12031-82-2
109.78
MFCD00016181

Specifications

Lithium titanium oxide
Powder
(metals basis)
MFCD00016181
Odorless
250g
12031-82-2
99%
109.78
−50 Mesh
1520° to 1564°C
Videos
SDS