Alfa Aesar™ Lithium titanium oxide, 99% (metals basis)


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 041650.A1

LITHIUM TITANIUM OXIDE 99%1KG

UNSPSC: 12141746

Code: NEW

Additional Details:
Additional Details: CAS Number: 12031-82-2 Weight: 0.01000kg


Quantity 1kg

Product Code. 11378788

Quantity Price
1 £ 527.46 / 1kg
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Lithium titanium oxide
CAS 12031-82-2
Formula Weight 109.78
MDL Number MFCD00016181

Specification

Formula Weight 109.78
Chemical Name or Material Lithium titanium oxide
TSCA TSCA
Mesh Size −50 Mesh
Odor Odorless
Physical Form Powder
Assay 99%
Assay Percent Notes (metals basis)
MDL Number MFCD00016181
CAS 12031-82-2
Melting Point 1520° to 1564°C
Quantity 1kg