missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetramethylcyclopentadienide, 97+%, Thermo Scientific™

Catalog Number 15403797
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
82061-21-0
C9H13Li
MFCD00269824
tetramethylcyclopentadienyl lithium, hsqylejmiwejbm-uhfffaoysa-n, 1,2,3,4-tetramethylcyclopentadienyl lithium, 2,3,4,5-tetramethylcyclopentadienyl lithium, lithium 1,2,3,4-tetramethyl-cyclopentadienide
This item is not returnable. View return policy
82061-21-0
C9H13Li
MFCD00269824
tetramethylcyclopentadienyl lithium, hsqylejmiwejbm-uhfffaoysa-n, 1,2,3,4-tetramethylcyclopentadienyl lithium, 2,3,4,5-tetramethylcyclopentadienyl lithium, lithium 1,2,3,4-tetramethyl-cyclopentadienide

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point >350°C
Quantity 5 g
UN Number UN3391
Sensitivity Air and moisture sensitive
Formula Weight 128.14
Percent Purity ≥97%
Chemical Name or Material Lithium tetramethylcyclopentadienide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'