missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-44357
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
12007-60-2
B4Li2O7
169.11
MFCD00011083
RDZCISUGRQUANA-UHFFFAOYSA-N
dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
12007-60-2
B4Li2O7
169.11
MFCD00011083
RDZCISUGRQUANA-UHFFFAOYSA-N
dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 930.0°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98+%
Linear Formula Li2B4O7
Merck Index 15, 5580
Solubility Information Solubility in water: 28g/L (20°C). Other solubilities: 52g/L (100°C)
Formula Weight 169.12
Percent Purity ≥98%
Grade Analytical
Loss on Drying 0.5% max. (200°C)
Packaging Plastic bottle
Chemical Name or Material Lithium tetraborate, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Lithium tetraborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'