missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals
132.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
800 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 800 mL |
Linear Formula | LiOCH(CH3)3 |
Flash Point | -11°C |
Formula Weight | 80.05 |
Specific Gravity | 0.86 |
Density | 0.8600g/mL |
Chemical Name or Material | Lithium tert-butoxide |
Safety and Handling
Product Identifier
- Lithium tert-butoxide, 8% in 2-MeTHF
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'