missing translation for 'onlineSavingsMsg'
Learn More

Lithium tert-butoxide, 99%, Thermo Scientific Chemicals

Catalog Number p-7080090
Click to view available options
Quantity:
250 g
1 kg
1907-33-1
C4H9LiO
80.055
MFCD00050479
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]
1907-33-1
C4H9LiO
80.055
MFCD00050479
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
23664764
lithium;2-methylpropan-2-olate
[Li+].CC(C)(C)[O-]

Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼150°C (decomposition)
Color White to Cream
Odor Odorless
Linear Formula (CH3)3COLi
Quantity 250 g
UN Number UN3206
Beilstein 3620018
Sensitivity Air and moisture sensitive
Solubility Information Soluble in toluene,hexane,tetrahydrofuran and methyl tert-butyl ether.
Formula Weight 80.06
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Lithium tert-butoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'