Learn More
Thermo Scientific Alfa Aesar Lithium tantalum tert-butoxide, 99.9% (metals basis), 10% w/v in tert-butanol
Brand: Thermo Scientific Alfa Aesar 036574.AC
Lithium tantalum tert-butoxide, 99.9% (metals basis), 10% w/v in tert-butanol
Code : NEW
Additional Details : CAS Number : 21864-30-2 Weight : 0.01000kg Transport : UN number : 1120 Chem class : 3 Pack group : II