missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11307577
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
This item is not returnable. View return policy
12136-58-2
Li2S
45.94
MFCD00011085
GLNWILHOFOBOFD-UHFFFAOYSA-N
lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis
64734
[Li+].[Li+].[S--]
This item is not returnable. View return policy
12136-58-2
Li2S
45.94
MFCD00011085
GLNWILHOFOBOFD-UHFFFAOYSA-N
lithium sulfide, lithium sulfide li2s, lithium sulphide, anhydrous, li2s, zwdbutfcwlvlcq-uhfffaoysa-n, lithium sulfide trace metals basis
64734
[Li+].[Li+].[S--]

It act as a superconductor and widely used in lithium-sulfur batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 900°C to 975°C
Color Yellow
Physical Form Powder
Quantity 2 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information soluble in water and ethanol
IUPAC Name dilithium(1+) sulfanediide
Formula Weight 45.95
Odor Like rotten eggs
Sensitivity Moisture sensitive
Appearance Yellow powder
Density 1.66 g/cm3 at 20°C
Chemical Name or Material Lithium sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'