missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate monohydrate, 99+%, pure, Thermo Scientific Chemicals

Catalog Number p-3523481
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
10102-25-7
H2Li2O5S
127.95
MFCD00149766
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
dilithium;sulfate;hydrate
[Li+].[Li+].O.[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10102-25-7
H2Li2O5S
127.95
MFCD00149766
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
dilithium;sulfate;hydrate
[Li+].[Li+].O.[O-]S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 860.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99+%
Linear Formula LI2SO4·H2O
Merck Index 15, 5597
Solubility Information Solubility in water: 295.5g/L (20°C)
Formula Weight 127.96
Percent Purity ≥99%
Grade Pure
Loss on Ignition 13.5% to 15% (600°C, 2 hrs)
Packaging Plastic bottle
Chemical Name or Material Lithium sulfate monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium sulfate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'