missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate monohydrate, 99%, Thermo Scientific™

Catalog Number p-4813516
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
10102-25-7
H2Li2O5S
127.95
MFCD00149766
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
dilithium(1+) hydrate sulfate
[Li+].[Li+].O.[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10102-25-7
H2Li2O5S
127.95
MFCD00149766
RKGLUDFWIKNKMX-UHFFFAOYSA-L
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n
165815
dilithium(1+) hydrate sulfate
[Li+].[Li+].O.[O-]S([O-])(=O)=O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 860.0°C
Quantity 100 g
Assay Percent Range 99%
Linear Formula Li2SO4·H2O
Solubility Information Solubility in water: 295.5g/L (20°C).
Formula Weight 127.96
Percent Purity 99%
Grade Biochemical
Packaging Plastic bottle
Chemical Name or Material Lithium sulfate monohydrate
compliance-icons
Product Identifier
  • Lithium sulfate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'