missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium sulfate monohydrate, 99%, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Melting Point | 860.0°C |
Quantity | 500 g |
Assay Percent Range | 99% |
Linear Formula | Li2SO4·H2O |
Solubility Information | Solubility in water: 295.5g/L (20°C). |
Formula Weight | 127.96 |
Percent Purity | 99% |
Grade | Biochemical |
Packaging | Plastic bottle |
Chemical Name or Material | Lithium sulfate monohydrate |
Safety and Handling
Product Identifier
- Lithium sulfate monohydrate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'