missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 859.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Assay Percent Range | 99% |
Linear Formula | Li2SO4 |
Merck Index | 15,5597 |
Solubility Information | Solubility in water: 25.7 g/100mL (25°C). Other solubilities: practically insoluble in alcohol |
IUPAC Name | dilithium(1+) sulfate |
Formula Weight | 109.94 |
Show More |
Safety and Handling
Product Identifier
- Lithium sulfate, anhydrous
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'