missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium stearate, Thermo Scientific Chemicals
27.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Lithium stearate is used as a thickener and gelling agent to prepare oils into lubricating greases. It is the active component of lithium grease. Further, it is used as lubricants, lubricant additives, plastic and rubber products. In addition to this, it is used as a stabilizing agent in the plastics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 220°C |
Molecular Formula | C9H11ClN3 |
Quantity | 250 g |
Synonym | 2-1h-benzo d imidazol-2-yl ethanamine dihydrochloride, 2-1h-benzimidazol-2-yl ethanamine dihydrochloride, 1h-benzimidazole-2-ethanamine dihydrochloride, 2-2-aminoethyl benzoimidazole dihydrochloride, 2-1h-benzoimidazol-2-yl ethylamine dihydrochloride, 1h-benzimidazole-2-ethanamine, dihydrochloride, 1h-benzimidazole-ethanamine dihydrochloride, 2-2-aminoethyl benzoimidazoledihydrochloride, 2-1h-1,3-benzodiazol-2-yl ethanamine dihydrochloride, 2-benzimidazol-2-ylethylamine, chloride, chloride |
Solubility Information | Insoluble in water. |
InChI Key | VLIBSZCLVYXJTG-UHFFFAOYSA-N |
SMILES | C1=CC=C2C(=C1)NC(=N2)CCN.Cl.Cl |
IUPAC Name | 2-(1H-benzimidazol-2-yl)ethanamine;dihydrochloride |
Molecular Weight (g/mol) | 196.66 |
PubChem CID | 2788071 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'