missing translation for 'onlineSavingsMsg'
Learn More

Lithium Standard for ICP, ICP-MS 10,000 µg/mL (10,000 ppm) in 2-5% Nitric Acid (HNO3), Reagecon

Manufactured from ultra-pure metal (99.999%) tested by an ISO17025 Accredited Test Method (INAB Ref: 264T) and raw materials assayed by titration and ICP-MS prior to manufacture.

Brand:  Reagecon PLI4B2

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 1760 Chem class : 8 Pack group : III

Product Code. 11488959

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • ICP, ICP-MS Reference Standard, ICP, ICP-MS Reference Materia
  • ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
  • ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
  • ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
  • ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
  • ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
  • GHS/OSHA compliant, REACH etc.
  • This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple leached pre-cleaned bottles.
  • The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
  • All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
Specifications

Specifications

Liquid
HDPE Triple-leached
250 mL
ICP, ICP-MS, AA, IC, XRF and other cation measurement techniques
10,000 µg/mL (10,000 ppm)
Lithium
Matrix: 2-5% HNO3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers