missing translation for 'onlineSavingsMsg'
Learn More

Lithium phosphate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048520
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10377-52-3
Li3O4P
115.79
MFCD00016187
TWQULNDIKKJZPH-UHFFFAOYSA-K
lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
165867
[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10377-52-3
Li3O4P
115.79
MFCD00016187
TWQULNDIKKJZPH-UHFFFAOYSA-K
lithium phosphate, lithium orthophosphate, trilithium orthophosphate, trilithium phosphate, phosphoric acid, trilithium salt, unii-2qm4k05q74, lithiumphosphate, trilithotab phosphate, phosphoric acid, lithium salt 1:3, li3po4
165867
[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O

Lithium phosphate batteries batteries are now widely used as a staple for cell phones, laptops and some other gadgets. Used in low-expansion porcelain enamel glazes and polymer intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 837°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Poor solubility in water. Insoluble in alcohols.
IUPAC Name trilithium(1+) phosphate
Formula Weight 115.79
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Density 2.54
Assay (metals basis)
Chemical Name or Material Lithium phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'