missing translation for 'onlineSavingsMsg'
Learn More

Lithium peroxide 95%, ACROS Organics™

Brand:  Acros Organics 389882500

Code : 29

Additional Details : CAS Number : 12031-80-0 Weight : 0.30000kg Transport : UN number : 1472 Chem class : 1 Pack group : II


Quantity 250g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10793245

  • £109.60 / 250g


Estimated Shipment
13-03-2018
Add to basket


Specifications

Chemical Identifiers

Lithium peroxide
95%
LiO2Li
MFCD00016184
≥94% (permanganate titration)
Li2O2
45.88

Specifications

Lithium peroxide
Powder
95%
LiO2Li
MFCD00016184
White to pale yellow
12031-80-0
≥94% (permanganate titration)
Li2O2
45.88
Plastic bottle
250g
Videos
SDS