missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium peroxide, 95%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Plastic Bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | White to Yellow |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 95% |
Linear Formula | LiO2Li |
Formula Weight | 45.88 |
Percent Purity | 95% |
Packaging | Plastic Bottle |
Chemical Name or Material | Lithium peroxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'