missing translation for 'onlineSavingsMsg'
Learn More

Lithium peroxide, 95%, Thermo Scientific Chemicals

Catalog Number 11316758
204.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12031-80-0
Li2O2
MFCD00016184
HPGPEWYJWRWDTP-UHFFFAOYSA-N
25489
dilithium;peroxide
[Li+].[Li+].[O-][O-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12031-80-0
Li2O2
MFCD00016184
HPGPEWYJWRWDTP-UHFFFAOYSA-N
25489
dilithium;peroxide
[Li+].[Li+].[O-][O-]

Lithium peroxide is used in air purifiers. It is also used to produce high-purity oxygen. It acts as a catalyst for polymerization of styrene to polystyrene. Further, it serves as a curing agent for special polymers and a source of oxygen in sealed spaces such as submarines and in breathing apparatus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 100 g
UN Number UN1472
Synonym dilithium peroxide, lithium peroxide li2 o2, lithium oxide, unii-9anx556r5f, lithium peroxide un1472 oxidizer
Solubility Information Soluble in anhydrous acetic acid. Insoluble in alcohol.
Molecular Weight (g/mol) 45.878
Formula Weight 45.88
Percent Purity 95%
Odor Odorless
Sensitivity Air and moisture sensitive
Chemical Name or Material Lithium peroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'