missing translation for 'onlineSavingsMsg'
Learn More

Lithium perchlorate trihydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number 10582981
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
13453-78-6
ClLiO4·3H2O
160.44
MFCD00149765
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
23708903
lithium;perchlorate;trihydrate
[Li+].O.O.O.[O-]Cl(=O)(=O)=O
This item is not returnable. View return policy
13453-78-6
ClLiO4·3H2O
160.44
MFCD00149765
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
23708903
lithium;perchlorate;trihydrate
[Li+].O.O.O.[O-]Cl(=O)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 95°C
Color White
pH 6.0 to 7.5 (5% soln. at 20°C)
Physical Form Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula LiClO4·3H2O
Merck Index 15, 5595
Solubility Information Solubility in water: 597g/L water (25°C). Other solubilities: soluble in alcohol
Formula Weight 160.44
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Lithium perchlorate trihydrate, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium perchlorate trihydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'