missing translation for 'onlineSavingsMsg'
Learn More

Lithium oxalate, 99+%, Thermo Scientific Chemicals

Catalog Number p-7049228
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
553-91-3
C2Li2O4
101.90
MFCD00040596
YNQRWVCLAIUHHI-UHFFFAOYSA-L
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
68383
[Li+].[Li+].[O-]C(=O)C([O-])=O
This item is not returnable. View return policy
553-91-3
C2Li2O4
101.90
MFCD00040596
YNQRWVCLAIUHHI-UHFFFAOYSA-L
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
68383
[Li+].[Li+].[O-]C(=O)C([O-])=O

Lithium oxalate is used as lithium battery electrolyte. It is also used as an analytical reagent and reducing reagent. Further, it is used as an intermediate in the manufacture of other chemicals such as pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Density 2.12 g/mL
Quantity 50 g
UN Number UN3288
Merck Index 14,5537
Solubility Information Soluble in water.
IUPAC Name dilithium;oxalate
Formula Weight 101.96
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Lithium oxalate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'