missing translation for 'onlineSavingsMsg'
Learn More

Lithium nitrate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog Number p-4407580
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
7790-69-4
LiNO3
68.94
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
[Li+].[O-][N+]([O-])=O
7790-69-4
LiNO3
68.94
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
[Li+].[O-][N+]([O-])=O

Used in Ceramics, Pyrotechnics, Salt Baths

Used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, and rocket propellant Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent in the production of red-colored fireworks and flares; as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from the sun for cooking. It is applied to concrete pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 600°C (decomposition)
Melting Point 264°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99%
Solubility Information Soluble in water,alcohol,ethanol,methanol,pyridine,ammonia and acetone.
IUPAC Name lithium;nitrate
Formula Weight 68.95
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Density 2.38 g/cm3 at 20°C
Chemical Name or Material Lithium nitrate, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'