missing translation for 'onlineSavingsMsg'
Learn More

Lithium nitrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3734863
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
7790-69-4
LiNO3
68.94
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
[Li+].[O-][N+]([O-])=O
7790-69-4
LiNO3
68.94
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
[Li+].[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 600.0°C
Melting Point 251.0°C
Color White
Physical Form Adhering Crystalline Powder
Quantity 250 g
Assay Percent Range 99+%
Linear Formula LiNO3
Merck Index 15, 5592
Solubility Information Solubility in water: 90g/L (28°C). Other solubilities: soluble in alcohol
IUPAC Name lithium;nitrate
Formula Weight 68.95
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 2.38
Density 2.3800g/mL
Chemical Name or Material Lithium nitrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'