missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
166.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Specifications
Boiling Point | 600.0°C |
Melting Point | 251.0°C |
Color | Black to Blue |
Physical Form | Lustrous Spheres |
Quantity | 100 g |
Assay Percent Range | 99.999% |
Linear Formula | LiNO3 |
Merck Index | 15,5592 |
Solubility Information | Solubility in water: 90g/L (28°C). Other solubilities: soluble in alcohol |
IUPAC Name | lithium;nitrate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'