missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium niobium methoxide, 99+% (metals basis), 5% w/v in methanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
Used as a precursor for chemical vapor deposition of thin films for lithium ion battery electrolyte materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Linear Formula | LiNb(OCH3)6 |
Quantity | 25 mL |
UN Number | UN1230 |
Sensitivity | Moisture sensitive |
Solubility Information | Fully miscible with water. |
Formula Weight | 285.92 |
Percent Purity | ≥99% |
Physical Form | Liquid |
Assay | (metals basis) |
Chemical Name or Material | Lithium niobium methoxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'