missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium metaborate, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 845°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Soluble in ethanol. Partially soluble in cold water. |
IUPAC Name | tetraboron heptahydrate dilithium |
Formula Weight | 49.75 |
Sensitivity | Hygroscopic |
Density | 1.397 |
Chemical Name or Material | Lithium metaborate, anhydrous |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'