missing translation for 'onlineSavingsMsg'
Learn More

Lithium metaborate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog Number p-7050415
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
This item is not returnable. View return policy
13453-69-5
B4H14Li2O7
183.23
MFCD00011089
DPYKRXYVXDYLEY-UHFFFAOYSA-N
lithium metaborate, boric acid hbo2 , lithium salt, lithium metaborate, anhydrous, lithium oxido oxo borane, bo2.li, boric acid hbo2 , lithium salt 1:1, lithium metaborate,anhydrous, lithium metaborate, puratronic, lithotab boron oxide hydroxide, lithium metaborate, acs
123308
[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O

It acts as a support for antioxidant property in human and animal tissues. Used in non-linear optic materials and cellular phones. It is used for measuring total elements in soil, geological, plant and other natural organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 845°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 98.0-102.0%
Linear Formula LiBO2
Solubility Information soluble in ethanol.
IUPAC Name tetraboron heptahydrate dilithium
Formula Weight 49.75
Percent Purity 98.0 to 102.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 1.397
Chemical Name or Material Lithium metaborate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'