missing translation for 'onlineSavingsMsg'
Learn More

Lithium metaborate, 99%, for analysis, ACROS Organics™

Brand:  Acros Organics 365715000

Code : 29

Additional Details : CAS Number : 13453-69-5 Weight : 0.55000kg

Packaging Plastic bottle
Quantity 500g

 View more versions of this product

Product Code. 10661653

  • £196.90 / 500g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

Lithium metaborate
13453-69-5
≥98.5%
BLiO2
49.75
99%
For analysis
99%
LiBO
MFCD00011089

Specifications

Lithium metaborate
13453-69-5
Powder
99%
LiBO
MFCD00011089
50ppm max.
Plastic bottle
1000ppm max.
100ppm max.
500g
99%
For analysis
≥98.5%
BLiO2
49.75
200ppm max.
100ppm max.
50ppm max.
100ppm max.
White
Videos
SDS